Caplugs™ (Evergreen Scientific) Sterile Polystyrene Culture Tubes
Precision molded from premium-grade, chemically inert plastic materials
Supplier:  Caplugs™ (Evergreen Scientific) 2222070010
 View more versions of this product
Catalog No. 22-047-505
Explore available promotions
Description
Description
  • Gamma radiation sterilized; shipped ready to use
  • Tubes are constructed of polystyrene for clarity
  • Polystyrene tubes may be centrifuged to 3000 × g
  • Overfitting caps have inner ribs for two-position fits: loose for aerobic culturing and tight for anaerobic culturing and sealing
  • Tabs and/or rings allow easy removal and sealing
  • Screw caps have built-in sealing rings to provide leakproof closures
  • Available with or without caps; packaged in easy-to-open bags of 25 tubes each
  • Individual packaging available for 12 × 75mm, 13 × 100mm, 17 × 100mm, and 16 × 125mm sizes (see Cat. Nos. 22-047-503, -509, and -507)
Specifications
Specifications
Cell Culture Tube
3000 x g
100 mm
500/Cs.
Sterile
No
17 mm
Dual-position Cap
Polystyrene
20 Bags of 25
16 mL