missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ansell MICROFLEX™ Evolution One™ EV-2050 Latex Gloves
Powder free, latex examination gloves provide maximum comfort for long shifts. Ideal for the life science and healthcare industries.
$16.65 - $16.65
Specifications
Certifications/Compliance | ASTM D3578, FDA21 CFR 177-2600 |
---|---|
Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
Chemotherapeutic Rated | No |
Applications | Laboratory, Healthcare |
AQL | 1.5 |
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
11-462-68B
|
Ansell
EV2050S |
Small |
Pack of 100 Each for $16.65
Case of 10 Pack for $166.50
|
|
|||||
11-462-68C
|
Ansell
EV2050M |
Medium |
Pack of 100 Each for $16.65
Case of 10 Pack for $166.50
|
|
|||||
11-462-68D
|
Ansell
EV2050L |
Large |
Pack of 100 Each for $16.65
Case of 10 Pack for $166.50
|
|
|||||
11-462-68E
|
Ansell
EV2050XL |
X-Large |
Pack of 100 Each for $16.65
Case of 10 Pack for $166.50
|
|
|||||
Description
Ideal for handling potentially hazardous materials.
- Tactile sensitivity and high tensile strength
- Textured for better handling in wet conditions
- Beaded cuffs for easy donning and doffing
- Ambidextrous, non-sterile, single-use
- Length: 9.6 in. (24.4 cm)
- Natural color
Specifications
ASTM D3578, FDA21 CFR 177-2600 | |
No | |
1.5 | |
Single-use | |
5.9 mil | |
No | |
Latex | |
0.14 mm | |
5.9 mil | |
Yes | |
25 MPa | |
24.4 cm |
EN 420:2003 + A1:2009 | |
Laboratory, Healthcare | |
Natural | |
Fully Textured | |
0.15 mm | |
No | |
5.5 mil | |
Powder Free | |
0.15 mm | |
Evolution One™ EV-2050 | |
9.6 in. |