missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

OHAUS™ Explorer™ Precision Balances

The Explorer has risen to the next level of ingenuity; expanding on its modern features and design to offer unmatched functionality.

Supplier:  OHAUS™ 30061981

 View more versions of this product

Catalog No. 02-112-241


Explore available promotions

Description

Description

  • Large color touch-screen display
  • Icon-driven application software that features 14 unique application modes
  • 3 level adjustable angle display
  • Newly optimized linearity and repeatability specifications 
  • Enhanced vibration filtering for better balance stability
  • All models come standard with AutoCal™ automatic internal calibration
  • 3 x 3 filtering setup ensures fast and stable weighing results
  • The base and display separate for maximum flexibility in anywork area 
  • Draftshield models isolate the weighing chamber to enhance stability and speed results while offering maximum accessibility to the weighing chamber through top-mounted side doors or through the versatile top door that flips or slides open
  • Hands-free entry is available with optional automated side doors
  • Easy access communication ports (USB, RS232) plus 3rd communication port slot (Ethernet or 2nd RS232)
  • GLP and GMP including date and time, Direct Data Transfer
  • Metal base, ABS top housing, stainless steel pan, glass draftshield with flip-top door, replaceable in-use cover 
  • 3-door draftshield with removable panels on 0.1mg and 1 mg models with draftshield light
  • Modular display design with adjustable angle (attach to base, wall or optional tower)
  • AutoCal automatic internalcalibration
  • Level assist feature with illuminated level indicator
  • Selectable environmental and auto-print settings
  • Integral weigh belowhook
  • Protective in-use cover
  • Software lock switch 
  • High resolution 14.5cm (diagonal) full colour VGA touch display including QWERTY and numeric key pads plus weighing capacity indication
 
Applications
Weighing (17 units + custom units), parts counting, percent weighing, animal/dynamic weighing, checkweighing, filling,totalization, formulation, differential weighing, density determination, peak hold, ingredient costing, pipette adjustment, gross/net/tare weighing
Specifications

Specifications

Touch screen
RS-232, USB, Optional Ethernet or 2nd RS232
0.01 g
Automatic draft shield 3 door, internal (AutoCal™)
ABS Plastic, Stainless Steel, Glass
<±0.01 g
1.5ppm/°C
50°F to 86°F
Precision Balance
23 cm
9.8 cm
39.3 cm
4.3 kg
100/240 V
50/60 Hz
NIST, AM-5847, IEC/EN 61010-1:2010, CAN/CSA-C22.2,
Color Graphic
Gram, milligram, kilogram, carat, ounce, troy ounc
±0.02 g
EX2202/E
2.2 kg
≤1 sec.
10°C to 30°C
9.1 in.
3.9 in.
15.5 in.
9.5 lb.
100/240 V 50/60 Hz
0.6 A
Videos
SDS
Documents

Documents

Promotions

Promotions