Falcon™ Chambered Cell Culture Slides
Packaged in disposable incubation trays. Falcon™ Chambered Cell Culture Slides are ideal for in situ DNA amplification/hybridization, tissue and tumor typing/pathology, or any study requiring a small, accessible culture area.
Supplier:  Falcon™ 354108
 View more versions of this product
Catalog No. 08774208
Explore available promotions
Description
Description
  • Simplify analysis: culture, fix, stain and  examine your samples under a microscope without cell transfer
  • Polystyrene chamber affixed to a specially cleaned and triple-rinsed glass microscope slide (25 × 75mm)
  • Safety Removal Tool separates wells from slide; gasket remains with plastic vessel, prevents cross-contamination and doesn't interfere with further processing
  • Sterile
  • Culture up to 8 different conditions on a single glass microscope slide
  • Numeric labeling; 4-well, and 8-well formats available
Specifications
Specifications
Cell Culture Slide
0.7 cm2
In situ DNA amplification or hybridization, tissue and tumor typing or pathology, or any study requiring a small, accessible culture area
96
8
0.7 to 0.8 mL
Handled polystyrene lid, safety removal tool
Sterile