missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Falcon™ Chambered Cell Culture Slides

Packaged in disposable incubation trays. Falcon™ Chambered Cell Culture Slides are ideal for in situ DNA amplification/hybridization, tissue and tumor typing/pathology, or any study requiring a small, accessible culture area.

$523.71 - $2025.26

Specifications

Type Cell Culture Slide
For Use With (Application) In situ DNA amplification or hybridization, tissue and tumor typing or pathology, or any study requiring a small, accessible culture area
Includes Handled polystyrene lid, safety removal tool
Sterility Sterile
View More Specs

Includes: Handled polystyrene lid, safety removal tool

Products 4
Catalog Number Mfr. No. Culture Area Wells No. per Case Volume (Metric) Working Price Quantity  
Catalog Number Mfr. No. Culture Area Wells No. per Case Volume (Metric) Working Price Quantity  
08774208
View Documents
Falcon™
354108
0.7 cm2 8 96 0.7 to 0.8 mL
Case of 96 Each for $2,025.26
 
08-774-209
View Documents
Falcon™
354104
1.7 cm2 4 96 1 to 1.5 mL
Case of 96 Each for $1,708.23
 
08-774-25
View Documents
Falcon™
354114
1.7 cm2 4 24 1 to 1.5 mL
Case of 24 Each for $534.21
 
08-774-26
View Documents
Falcon™
354118
0.7 cm2 8 24 0.7 to 0.8 mL
Case of 24 Each for $523.71
 
Description

Description

 

  • Simplify analysis: culture, fix, stain and  examine your samples under a microscope without cell transfer
  • Polystyrene chamber affixed to a specially cleaned and triple-rinsed glass microscope slide (25 × 75mm)
  • Safety Removal Tool separates wells from slide; gasket remains with plastic vessel, prevents cross-contamination and doesn't interfere with further processing
  • Sterile
  • Culture up to 8 different conditions on a single glass microscope slide
  • Numeric labeling; 4-well, and 8-well formats available

Order Info

Available with 1, 2, 4, or 8 wells, numbered for easy identification. Supplied with handled polystyrene lid. Packaged in disposable incubation trays. Each tray holds 12 slides.

Specifications

Specifications

Cell Culture Slide
In situ DNA amplification or hybridization, tissue and tumor typing or pathology, or any study requiring a small, accessible culture area
Handled polystyrene lid, safety removal tool
Sterile
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'