missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Falcon™ Standard Tissue Culture Dishes
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
Supplier: Falcon™ 353003
Description
- Flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells
- Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment
- Improved handling of small dishes with the unique Easy-Grip design
- Nonpyrogenic
- Sterilized by gamma irradiation
- Packaged in peel-open, medical-style bags
- Crystal-grade virgin polystyrene
- Standard Tissue Culture (TC), Corning™ PrimariaTM surface treatment and non-TC-treated polystyrene available
Designed for cell culture
- Flat, distortion-free optics
- Lids designed for optimal gas exchange
- Stacking rings allow for easier stacking and handling
- Vacuum-gas plasma treatment permanently and consistently modifies the cell growth surface
- Standard tissue culture surface is hydrophilic and contains a variety of negatively charged functional groups that support cell attachment and spreading
- Corning Primaria tissue culture surface additionally incorporates nitrogen-containing functional groups and has been shown to support improved attachment, spreading, and differentiation of some cell types
Specifications
| Flat | |
| Clear | |
| Polystyrene | |
| Sterile | |
| Cell Culture Petri Dish | |
| 200/Cs. | |
| 20 mm |
| 17.5 mL (Well) | |
| 58.1 cm2 | |
| Round | |
| Cell Culture | |
| 16 to 17.5 mL | |
| 20/Pk. | |
| 100 mm |