Falcon™ Standard Tissue Culture Dishes
Constructed of flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells. Falcon™ Standard Tissue Culture Dishes works with most attachment-dependent cell lines.
$627.09 - $725.79
Specifications
Bottom Shape Flat
Color Clear
Material Polystyrene
Shape Round
Sterility Sterile
View More Specs
Products 2
Catalog Number Mfr. No. Volume (Metric) Working Culture Area Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Working Culture Area Price Quantity  
08-772E
View Documents
Falcon™
353003
16 to 17.5 mL 58.1 cm2
Case of 200 Each for $627.09
08-772B
View Documents
Falcon™
353002
6 to 7 mL 21.29 cm2
Case of 500 Each for $725.79
Description
Description
  • Flat, optically clear polystyrene surfaces for distortion-free microscopic visualization of cells
  • Uniform surface chemistry created by vacuum-gas plasma treatment promotes cell attachment
  • Improved handling of small dishes with the unique Easy-Grip design
  • Nonpyrogenic
  • Sterilized by gamma irradiation
  • Packaged in peel-open, medical-style bags
  • Crystal-grade virgin polystyrene
  • Standard Tissue Culture (TC), Corning™ PrimariaTM surface treatment and non-TC-treated polystyrene available
Designed for cell culture
  • Flat, distortion-free optics
  • Lids designed for optimal gas exchange
  • Stacking rings allow for easier stacking and handling
  • Vacuum-gas plasma treatment permanently and consistently modifies the cell growth surface
  • Standard tissue culture surface is hydrophilic and contains a variety of negatively charged functional groups that support cell attachment and spreading
  • Corning Primaria tissue culture surface additionally incorporates nitrogen-containing functional groups and has been shown to support improved attachment, spreading, and differentiation of some cell types
Order Info
Packed 20 per sleeve.
Specifications
Specifications
Flat
Polystyrene
Sterile
Cell Culture Petri Dish
Clear
Round
Cell Culture
20/Pk.
SDS
missing translation for 'documents'
missing translation for 'documents'