missing translation for 'onlineSavingsMsg'
Learn More

Fehling B solution

CAS: 6381-59-5

Supplier:  Thermo Scientific Chemicals 270790010

 View more versions of this product

Catalog No. AC270790010

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Fehling B solution
6381-59-5,1310-73-2,7732-18-5
MFCD00065391
Plastic bottle
1 L
Colorless
100.0°C
1.2400g/mL
1.24
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only