missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Specifications
Chemical Name or Material | Fehling B solution |
---|---|
Color | Colorless |
CAS | 6381-59-5,1310-73-2,7732-18-5 |
Boiling Point | 100.0°C |
MDL Number | MFCD00065391 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AC270790010
|
Thermo Scientific Chemicals
270790010 |
1 L |
Each for $187.49
|
|
||||||
AC270790025
|
Thermo Scientific Chemicals
270790025 |
2.5 L |
Each for $387.96
|
|
||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fehling B solution | |
6381-59-5,1310-73-2,7732-18-5 | |
MFCD00065391 | |
Plastic bottle | |
Liquid |
Colorless | |
100.0°C | |
1.2400g/mL | |
1.24 |
RUO – Research Use Only