missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ferric Ammonium Sulfate Dodecahydrate (Crystalline/Certified ACS), Fisher Chemical™
Supplier: Thermo Fisher Scientific I75500
Specifications
230°C | |
39°C | |
Purple | |
Solid | |
98.5 to 102.0 % | |
FeNH4(SO4)2·12H2O | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] | |
482.176 | |
482.2 | |
Certified ACS | |
Glass Bottle |
Ferric Ammonium Sulfate Dodecahydrate | |
7783-83-7 | |
2.5 | |
500 g | |
FeH28NO20S2 | |
MFCD00150004 | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
azanium;iron(3+);disulfate;dodecahydrate | |
197097 | |
98.5 to 102.0% | |
Pass test | |
1.710g/cm³ |
Chemical Identifiers
7783-83-7 | |
482.176 | |
LCPUDZUWZDSKMX-UHFFFAOYSA-K | |
197097 | |
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3] |
FeH28NO20S2 | |
MFCD00150004 | |
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g | |
azanium;iron(3+);disulfate;dodecahydrate |