missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Ammonium Sulfate Dodecahydrate (Crystalline/Certified ACS), Fisher Chemical™

$419.50 - $1518.67

Chemical Identifiers

CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
MDL Number MFCD00150004
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
I75-500
SDS View Documents
Thermo Fisher Scientific
I75500
500 g Glass Bottle
Each for $419.50
Case of 6 Each for $2,517.00
 
I75-3
SDS View Documents
Thermo Fisher Scientific
I753
3 kg Glass Bottle
Each for $1,518.67
Case of 4 Each for $6,074.68
 
Description

Description

Specifications

Chemical Identifiers

7783-83-7
482.176
LCPUDZUWZDSKMX-UHFFFAOYSA-K
197097
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
FeH28NO20S2
MFCD00150004
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
azanium;iron(3+);disulfate;dodecahydrate

Specifications

230°C
7783-83-7
2.5
500 g
FeH28NO20S2
MFCD00150004
LCPUDZUWZDSKMX-UHFFFAOYSA-K
azanium;iron(3+);disulfate;dodecahydrate
197097
98.5 to 102.0%
Pass test
1.710g/cm³
39°C
Purple
Solid
98.5 to 102.0 %
FeNH4(SO4)2·12H2O
ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
482.176
482.2
Certified ACS
Glass Bottle
Ferric Ammonium Sulfate Dodecahydrate
Videos
Safety and Handling

Safety and Handling

SDS
missing translation for 'documents'

missing translation for 'documents'