missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ferric Chloride Hexahydrate (Lumps/Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific I88500

 View more versions of this product

Catalog No. I88-500



Explore available promotions

Description

Description

Specifications

Specifications

280°C
37°C
Yellow
Solid
97 to 102 %
Cl3FeH12O6
1773
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
270.3
Certified ACS
Poly Bottle
1.82g/mL
Ferric Chloride Hexahydrate
10025-77-1
2
500 g
97.0-102.0%
MFCD00149712
Iron (III) Chloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
270.29
97 to 102%
Pass test
Negligible

Chemical Identifiers

10025-77-1
270.29
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
Cl3FeH12O6
MFCD00149712
Iron (III) Chloride
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Causes burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.

NFPA

Health:3
Flammability:0
Instability:1

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions