Learn More
Ferric Chloride Hexahydrate (Lumps/Certified ACS), Fisher Chemical™
$307.50 - $485.33
Chemical Identifiers
CAS | 10025-77-1 |
---|---|
Molecular Formula | Cl3FeH12O6 |
Molecular Weight (g/mol) | 270.29 |
MDL Number | MFCD00149712 |
InChI Key | NQXWGWZJXJUMQB-UHFFFAOYSA-K |
Synonym | Iron (III) Chloride |
IUPAC Name | iron(3+) hexahydrate trichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
I88-100
|
Thermo Fisher Scientific
I88100 |
100 g | Poly Bottle |
Each for $307.50
Case of 6 Each for $1,845.00
|
|
||||
I88-500
|
Thermo Fisher Scientific
I88500 |
500 g | Poly Bottle |
Each for $485.33
Case of 6 Each for $2,911.98
|
|
||||
Chemical Identifiers
10025-77-1 | |
270.29 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride |
Cl3FeH12O6 | |
MFCD00149712 | |
Iron (III) Chloride | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Specifications
280°C | |
10025-77-1 | |
2 | |
100 g | |
Cl3FeH12O6 | |
1773 | |
NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
iron(3+) hexahydrate trichloride | |
270.3 | |
Certified ACS | |
Poly Bottle | |
1.82g/mL | |
97.0-102.0% |
37°C | |
Yellow | |
Solid | |
97 to 102% | |
MFCD00149712 | |
Iron (III) Chloride | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
270.29 | |
97 to 102% | |
Pass test | |
Negligible | |
Ferric Chloride Hexahydrate |
Safety and Handling
DANGER!
Emergency Overview
Causes burns by all exposure routes. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. Immediate medical attention is required. Do not induce vomiting.
NFPA
Health:3
Flammability:0
Instability:1
DOTInformation : DOT Class 8, : Corrosive