missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, charcoal stripped, USDA-approved regions DFS Item

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays.

Supplier:  Gibco™ A3382101

 View more versions of this product

Catalog No. A3382101


Explore available promotions

Description

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Serum Treatment: Stripped with charcoal-dextran
  • Recommended Storage: -5°C to -20°C
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Bovine
Liquid
Fetal Bovine Serum
Charcoal-stripped
One Shot™
Fetal
United States or Mexico
One Shot
50 mL
Sterile
Dry Ice
Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.