missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Fetal Bovine Serum, charcoal stripped, USDA-approved regions
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays.
$157.00 - $1,246.00
Specifications
Age | Fetal |
---|---|
Country of Origin | United States or Mexico |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Serum Treatment | Charcoal-stripped |
Description
Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera Category: Specialty
- Serum Treatment: Stripped with charcoal-dextran
- Recommended Storage: -5°C to -20°C
Order Info
Shipping Condition: Dry Ice
Specifications
Fetal | |
Liquid | |
Charcoal-stripped |
United States or Mexico | |
Fetal Bovine Serum | |
Dry Ice |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.