missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, dialyzed, US origin DFS Item

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

Supplier:  Gibco™ A3382001

 View more versions of this product

Catalog No. A3382001


Explore available promotions

Description

Description

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Bovine
Liquid
Fetal Bovine Serum
Dialyzed
One Shot™
Fetal
United States
One Shot
50 mL
Sterile
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.