missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Fetal Bovine Serum, dialyzed, US origin
Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.
$213.00 - $1618.00
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Age | Fetal |
Country of Origin | United States |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Description
Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements
- Sera category: Specialty
- Origin: United States
- Endotoxin level: ≤50 EU/mL (Specification is checked and recorded; levels routinely ≤ 10 EU/mL)
- Hemoglobin level: ≤25 mg/dL
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
United States | |
Fetal Bovine Serum | |
Frozen |
Fetal | |
Liquid | |
Dialyzed |
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.