missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum (FBS), Premium

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Supplier:  Gibco™ A5670701

 View more versions of this product

Catalog No. A5670701


Explore available promotions

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <10 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Specifications

Specifications

Store at ≤-10°C.
Cattle/Bovine
Frozen
Fetal Bovine Serum
Sterile-filtered
Premium
≤10 EU/mL
≤25 mg/dL
Dry Ice
Fetal
Refer to CoA for specific origin
Bottle
500 mL
Triple-filtered, 0.1 μm
Up to 60 quality tests including 9CFR virus testing, endotoxin, and performance
Less waste
Premium
SDS
Documents

Documents

Promotions

Promotions