missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, Canada DFS Item

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

Supplier:  Gibco™ 12484028

 View more versions of this product

Catalog No. 12484028


Explore available promotions

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Value (Standard)
  • Origin: Canada
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Bottle
500 mL
Research Grade, Qualified
Fetal
Canada
Fetal Bovine Serum
Heat-inactivated
Frozen
Videos
SDS
Documents

Documents

Promotions

Promotions