missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Fetal Bovine Serum, qualified, heat inactivated, Canada DFS Item

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

$780.00 - $780.00

Specifications

Content And Storage Storage conditions: ≤-10°C
Shipping conditions: Frozen
Age Fetal
Species Cattle/Bovine
Country of Origin Canada
Format Bottle
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
12484028 DFS Item
View Documents
Gibco™
12484028
500 mL
Each for $780.00
 
Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

  • Sera Category: Value (Standard)
  • Origin: Canada
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Bottle
Heat-inactivated
Frozen
Fetal
Canada
Fetal Bovine Serum
Research Grade, Qualified
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'