Thermo Scientific™ Finntip™ Stepper Pipette Tips
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Supplier:  Thermo Scientific™ 9404190
 View more versions of this product
Catalog No. 21-377-152
Explore available promotions
Description
Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
  • Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Specifications
2.5 mL
Non-filtered
ISO 9001, ISO 14001
Thermo Scientific FinnPipette Stepper
50, 100, 150, 200, 250μL
Non-sterile
Bulk
FinnTip