Thermo Scientific™ Finntip™ Stepper Pipette Tips
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
$32.96 - $462.74
Specifications
Filtered | Non-filtered |
---|---|
Certifications/Compliance | ISO 9001, ISO 14001 |
For Use With (Equipment) | Thermo Scientific FinnPipette Stepper |
Product Line | FinnTip |
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Volume (Metric) Dispensing | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Volume (Metric) | Sterility | Volume (Metric) Dispensing | Price | Quantity | |||
21-377-148
|
Thermo Scientific™
9404170 |
500 μL | Non-sterile | 10, 20, 30, 40, 50μL |
Pack of 100 Each for $462.74
|
|
|||
21-377-149
|
Thermo Scientific™
9404173 |
500 μL | Sterile | 10, 20, 30, 40, 50μL |
Pack of 50 Each for $314.78
|
|
|||
21-377-150
|
Thermo Scientific™
9404180 |
1250 μL | Non-sterile | 25, 50, 75, 100, 125μL |
Pack of 100 Each for $462.74
|
|
|||
21-377-151
|
Thermo Scientific™
9404183 |
1250 μL | Sterile | 25, 50, 75, 100, 125μL |
Pack of 50 Each for $32.96
|
|
|||
21-377-152
|
Thermo Scientific™
9404190 |
2.5 mL | Non-sterile | 50, 100, 150, 200, 250μL |
Pack of 100 Each for $263.12
|
|
|||
21-377-153
|
Thermo Scientific™
9404193 |
2.5 mL | Sterile | 50, 100, 150, 200, 250μL |
Pack of 50 Each for $178.75
|
|
|||
21-377-154
|
Thermo Scientific™
9404200 |
5 mL | Non-sterile | 100, 200, 300, 400, 500μL |
Pack of 50 Each for $235.48
|
|
|||
21-377-155
|
Thermo Scientific™
9404203 |
5 mL | Sterile | 100, 200, 300, 400, 500μL |
Pack of 25 Each for $157.61
|
|
|||
21-377-156
|
Thermo Scientific™
9404210 |
12.5 mL | Non-sterile | 250, 500, 750, 1000, 1250μL |
Pack of 50 Each for $232.49
|
|
|||
21-377-157
|
Thermo Scientific™
9404213 |
12.5 mL | Sterile | 250, 500, 750, 1000, 1250μL |
Pack of 25 Each for $152.57
|
|
|||
21-377-158
|
Thermo Scientific™
9404220 |
25 mL | Non-sterile | 500, 1000, 1500, 2000, 2500μL |
Pack of 20 Each for $101.33
|
|
|||
21-377-159
|
Thermo Scientific™
9404223 |
25 mL | Sterile | 500, 1000, 1500, 2000, 2500μL |
Pack of 10 Each for $42.47
|
|
|||
21-377-160
|
Thermo Scientific™
9404230 |
50 mL | Non-sterile | 1000, 2000, 3000, 4000, 5000μL |
Pack of 10 Each for $38.18
|
|
|||
21-377-161
|
Thermo Scientific™
9404233 |
50 mL | Sterile | 1000, 2000, 3000, 4000, 5000μL |
Pack of 10 Each for $72.76
|
|
|||
Description
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
- Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Non-filtered | |
ISO 9001, ISO 14001 | |
Thermo Scientific FinnPipette Stepper | |
FinnTip |