missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher BioReagents™ Microbiology Media: LB Broth, Miller

Used for growing molecular biology strains of E. coli-containing recombinant plasmids

Supplier:  Fisher BioReagents™ BP14262

 View more versions of this product

Catalog No. BP1426-2



Explore available promotions

Description

Description

Dehydrated.

Specifications

Specifications

4°C
Powder
LB Broth
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L
Poly Pail
Bacterial Culture
Molecular genetics
Microbiology Media
For preparing bacterial nutrient media
2000 g
Videos
SDS
Documents

Documents

Promotions

Promotions