missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher BioReagents™ Microbiology Media: LB Broth, Miller
Used for growing molecular biology strains of E. coli-containing recombinant plasmids
$174.98 - $536.93
Specifications
Content And Storage | 4°C |
---|---|
Form | Powder |
Grade | Molecular genetics |
Media Type | LB Broth |
Product Type | Microbiology Media |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
BP1426-2
|
Fisher BioReagents™
BP14262 |
2000 g |
Each for $536.93
Case of 4 Each for $2,147.72
|
|
||||||
BP1426-500
|
Fisher BioReagents™
BP1426500 |
500 g |
Each for $174.98
|
|
||||||
Description
Dehydrated.
Specifications
4°C | |
Molecular genetics | |
Microbiology Media | |
For preparing bacterial nutrient media |
Powder | |
LB Broth | |
Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 10g/L |