Fisher Chemical™ Permount™ Mounting Medium
For both mounting and long-term storage of slides
$281.83 - $621.50
Specifications
Applications Mounting and long-term storage of slides
Product Type Slide Mounting Media
Packaging Amber Glass
View More Specs
Alert:
This chemical is considered hazardous by the 2012 OSHA Hazard Communic. Standard (29 CFR 1910.1200).
Products 2
Catalog Number Mfr. No. Volume (English) Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Volume (English) Volume (Metric) Price Quantity  
SP15-100 Edge
View Documents
Fisher Chemical™
SP15100
3.38 oz. 100 mL
Each for $281.83
Case of 6 Each for $1,690.98
Edge
SP15-500 Edge
View Documents
Fisher Chemical™
SP15500
16.9 oz. 500 mL
Each for $621.50
Case of 6 Each for $3,729.00
Edge
Description
Description
For both mounting and long-term storage of slides
  • Low viscosity
  • Gives a thinner adhesive layer for better optical quality
  • A refractive index near that of fixed protein helps keep image free of optical imperfections such as diffraction halos
  • Suitable for mounting cover slips to slides with thick or thin specimens
  • Won't shift the colors of biological dyes
  • Won't lose its cohesiveness, adhesive quality, or color with age
  • Preserves most biological dyes with little or no fading when slides are stored in darkness
Note: Highly flammable liquid and vapor. May be fatal if swallowed and enters airways. Causes skin irritation. Causes serious eye irritation.
Specifications
Specifications
Mounting and long-term storage of slides
Slide Mounting Media
Amber Glass
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'