Fisher Chemical™ Permount™ Mounting Medium Edge
Edge
For both mounting and long-term storage of slides
Supplier:  Fisher Chemical™ SP15500
 View more versions of this product
Catalog No. SP15-500
Edge
Explore available promotions
Alert:
This chemical is considered hazardous by the 2012 OSHA Hazard Communic. Standard (29 CFR 1910.1200).
Description
Description
For both mounting and long-term storage of slides
  • Low viscosity
  • Gives a thinner adhesive layer for better optical quality
  • A refractive index near that of fixed protein helps keep image free of optical imperfections such as diffraction halos
  • Suitable for mounting cover slips to slides with thick or thin specimens
  • Won't shift the colors of biological dyes
  • Won't lose its cohesiveness, adhesive quality, or color with age
  • Preserves most biological dyes with little or no fading when slides are stored in darkness
Specifications
Specifications
Mounting and long-term storage of slides
Slide Mounting Media
500 mL
16.9 oz.
Amber Glass