Fisher Healthcare™ Pinnacle Portfolio Cyto-Stain™
Can be used for all gynecological and non-gynecological procedures
Supplier:  Fisher Healthcare™ 220103
 View more versions of this product
Catalog No. 22-220-103
Explore available promotions
Description
Description
Cyto-Stain Features:
  • Uniquely formulated cytoplasmic stain produces results similar to those of conventional Papanicolaou stains
  • Provides a full range of familiar cytoplasmic colors and allows the user control over staining intensities
  • Reduces staining time; cost effective because you need only one counterstain, not two
  • Staining characteristics are comparable to conventional EA-OG stains
  • Cyto-Stain G produces greener cyanophilic hues in intermediate and basal cells
Pinnacle Portfolio
  • Designed as a system
  • Each component has added value and benefits that reduce cost per slide
  • Unique chemical formulae of the promote long-term chemical stability, which extends the usable life of each component
  • Are subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity
Specifications
Specifications
Cytostain
Liquid
Can be used for all gynecological and non-gynecological procedures
4 x 500mL