missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Healthcare™ Pinnacle Portfolio Cyto-Stain™
Can be used for all gynecological and non-gynecological procedures
$142.56 - $142.56
Specifications
Type | Cytostain |
---|---|
Format | Liquid |
For Use With (Application) | Can be used for all gynecological and non-gynecological procedures |
Quantity | 4 x 500mL |
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
22-220-103
|
Fisher Healthcare™
220103 |
Cytostain |
Each for $142.56
Case of 4 Each for $570.24
|
|
|||||
Description
Cyto-Stain Features:
- Uniquely formulated cytoplasmic stain produces results similar to those of conventional Papanicolaou stains
- Provides a full range of familiar cytoplasmic colors and allows the user control over staining intensities
- Reduces staining time; cost effective because you need only one counterstain, not two
- Staining characteristics are comparable to conventional EA-OG stains
- Cyto-Stain G produces greener cyanophilic hues in intermediate and basal cells
Pinnacle Portfolio
- Designed as a system
- Each component has added value and benefits that reduce cost per slide
- Unique chemical formulae of the promote long-term chemical stability, which extends the usable life of each component
- Are subjected to some of the highest quality manufacturing standards in the industry
- Packaging improvements enhance product longevity
Specifications
Cytostain | |
Liquid | |
Can be used for all gynecological and non-gynecological procedures | |
4 x 500mL |