missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Healthcare™ PROTOCOL™ Gram Stain Sets
Reusable tray provides secure bottle storage
Supplier: Fisher Healthcare™ 2300583
Description
- Manufactured with traditional formulations used by most laboratories
- Complete stain sets provide convenient staining
- Decolorizer is acetone/isopropanol (1:4)
Order Info
All sizes supplied in polyethylene bottles; dispenser caps provided with the 8 oz. and 16 oz. sizes. Gallon-size Iodine is available as traditional diluent/concentrate or as ready-to-use Lugols.
Specifications
Gram Stain Kit | |
250mL gram crystal violet , 250mL stabilized iodine , 250mL decolorizer and 250mL safranin | |
Laboratory chemicals | |
Gram Stain |
1 Set | |
Bottle | |
Reddish Violet |