missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisher Healthcare™ PROTOCOL™ Gram Stain Sets

Reusable tray provides secure bottle storage

$483.84 - $483.84

Specifications

Type Gram Stain Kit
Quantity 1 Set
Format Bottle
For Use With (Application) Laboratory chemicals
Color Reddish Violet
View More Specs

Products 1
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
2300583
View Documents
Fisher Healthcare™
2300583
250mL gram crystal violet , 250mL stabilized iodine , 250mL decolorizer and 250mL safranin
Each for $483.84
 
Description

Description

  • Manufactured with traditional formulations used by most laboratories
  • Complete stain sets provide convenient staining
  • Decolorizer is acetone/isopropanol (1:4)

Order Info

All sizes supplied in polyethylene bottles; dispenser caps provided with the 8 oz. and 16 oz. sizes. Gallon-size Iodine is available as traditional diluent/concentrate or as ready-to-use Lugols.

Specifications

Specifications

Gram Stain Kit
Bottle
Reddish Violet
1 Set
Laboratory chemicals
Gram Stain
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'