missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Healthcare™ PROTOCOL™ Hema-Quik™ and Hema-Quik II Stain Solutions
Carefully balanced for optimum staining pH
Supplier: Fisher Healthcare™ 206441
Description
Stains for manual and automated staining methods.
- All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
- Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
- Remain stable for a minimum of 12 months when stored at room temperature
- Allow varied staining times to suit individual preferences
- Packaged in durable plastic bottles for safety and easy handling
- Convenient finger tabs and clear reagent identification eliminate improper line connection
Specifications
90mL | |
65°C | |
Hema-Quik II Stain Solution | |
Dark Blue |
Room Temperature | |
Liquid | |
For Wright giemsa staining, fast single solution manual staining |