missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisher Healthcare™ PROTOCOL™ Hema-Quik™ and Hema-Quik II Stain Solutions
Carefully balanced for optimum staining pH
$107.68 - $722.48
Specifications
Storage Requirements | Room Temperature |
---|---|
Boiling Point | 65°C |
Format | Liquid |
Color | Dark Blue |
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Quantity | Price | Quantity | ||||
23-123745
|
Fisher Healthcare™
123745 |
Hema-Quik Stain Solution | 1L |
Each for $251.45
Case of 12 Each for $3,017.40
|
|
||||
23-123919
|
Fisher Healthcare™
123919 |
Hema-Quik Stain Solution | 90mL |
Each for $186.90
Case of 27 Each for $5,046.30
|
|
||||
23-206441
|
Fisher Healthcare™
206441 |
Hema-Quik II Stain Solution | 90mL |
Pack for $107.68
Case of 27 Pack for $2,907.36
|
|
||||
23-200725
|
Fisher Healthcare™
200725 |
Hema-Quik II Stain Solution | 1L |
Each for $324.39
Case of 12 Each for $3,892.68
|
|
||||
23-250466
|
Fisher Healthcare™
250466 |
Hema-Quik Stain Solution | 1 gal. |
Each for $722.48
Case of 4 Each for $2,889.92
|
|
||||
Description
Stains for manual and automated staining methods.
- All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
- Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
- Remain stable for a minimum of 12 months when stored at room temperature
- Allow varied staining times to suit individual preferences
- Packaged in durable plastic bottles for safety and easy handling
- Convenient finger tabs and clear reagent identification eliminate improper line connection
Specifications
Room Temperature | |
65°C | |
Liquid | |
Dark Blue |