Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains
Manufactured with strict quality control to ensure lot-to-lot uniformity
Supplier:  Fisher Healthcare™ 245653
 View more versions of this product
Catalog No. 23-245653
Explore available promotions
Description
Description
Prepared with stain certified by the Biological Stain Commission
Specifications
Specifications
120°C
1X
Liquid
4L
Maroon
Gill Hematoxylin (1X)
Used for Histological/Cytological Staining