Fisher Healthcare™ PROTOCOL™ Histological/Cytological Stains
Manufactured with strict quality control to ensure lot-to-lot uniformity
Supplier:  Fisher Healthcare™ 245656
 View more versions of this product
Catalog No. 23-245656
Explore available promotions
Description
Description
Prepared with stain certified by the Biological Stain Commission
Specifications
Specifications
3X
Liquid
475mL
Gill Hematoxylin (3X)
Used for Histological/Cytological Staining