Fisherbrand™ Deck-Mounted 90° Swivel Eyewashes with Pop-Off Spray Head Covers
Space-saving design available in left or right mounts
Supplier:  Fisherbrand™ S19270BL
 View more versions of this product
Catalog No. 19-120-2309
Explore available promotions
Description
Description
Unit design saves space and fits easily into any work environment.
  • Two outlet heads mounted on chrome-plated brass supply arms
  • Hand operated by large, highly visible stainless-steel push handle
  • Pop-off dust covers protect sprayheads
  • Integral flow control assures safe, steady flow under water conditions from 30-90psi
  • With chrome-plated brass 0.5 in. (1.3cm) IPS stay-open ball valve
Specifications
Specifications
Push Handle
Unit Design saves Space and Fits easily into any Work Environment
Left Side
ANSI Z358.1-2004, SEI
Two outlet head, stay open Chrome Plated Brass IPS ball valve, pop-off dust cover, Stainless Steel push handle
90° Swivel Eyewash