Fisherbrand™ accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
For use on accumet™ Excel (XL) series benchtop meters.
Supplier: Fisherbrand™ 13620100
Includes: 8-pin connectors
Description
For use on accumet™ Excel (XL) series benchtop meters
- Immersion-type cell
- Use in standard electrode holder: unique compact size is same as typical pH electrode
- Built-in temperature sensors provide temperature-compensated readings
- Epoxy body
Specifications
1 cm-1 | |
AB200, XL20, XL200, XL30, XL50, XL500, XL60, XL600 Benchtop Meters | |
Epoxy | |
Immersed in deionized water |
Conductivity Probe | |
8-pin | |
10.0 to 2000 μS/cm | |
2 Cell Conductivity |