Fisherbrand™ accumet™ Temperature-Compensated Two-Cell Conductivity Probes, Mercury-Free
For use on accumet™ Excel (XL) series benchtop meters.
Supplier:  Fisherbrand™ 13620101
 View more versions of this product
Catalog No. 13-620-101
Explore available promotions
Includes: 8-pin connectors
Description
Description
For use on accumet™ Excel (XL) series benchtop meters
  • Immersion-type cell
  • Use in standard electrode holder: unique compact size is same as typical pH electrode
  • Built-in temperature sensors provide temperature-compensated readings
  • Epoxy body
Specifications
Specifications
0.1 cm-1
AB200, XL20, XL200, XL30, XL50, XL500, XL60, XL600 Benchtop Meters
Epoxy
Immersed in deionized water
Conductivity Probe
8-pin
0.5 to 200 μS/cm
2 Cell Conductivity