Fisherbrand™ Anti-Static Static-Away™ Handle Brushes
Anti-Static Static-Away™ Handle Brushes designed with the softest hairs available to prevent scratching.
 View more versions of this product
Catalog No. 15-078-214
Explore available promotions
Description
Description
Anti-Static Static-Away™ Handle Brushes designed with the softest hairs available to prevent scratching.
  • Remove static charges as they brush
  • Conductive composite handle
  • Long handles allow access to difficult places
  • Eliminate the possibility of a hand touching
  • Weight: <1 oz. (28.3 g)
Specifications
Specifications
Anti-static Brush
Anti-static
3 in.
7.62 cm
0.5 oz.
Soft
Ideal for dusting balances, cuvets, microscope slides, optics and instrument/computer screens, designed with the softest hairs available to prevent scratching
6 in.
15.24 cm
14.17 g