Fisherbrand™ Traceable™ Digital Thermometer with Stainless-Steel Stem, 0.25 in. LCD Screen, and Protective Guard
Easy-to-read handheld Traceable™ Digital Thermometer.
 View more versions of this product
Catalog No. 14-648-12
Explore available promotions
Description
Description
Easy-to-read handheld Traceable™ Digital Thermometer.
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading—ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
Certifications
Multi-point calibration on individually-numbered Traceable Certificate assures accuracy from ISO/IEC 17025:2005 (1750.01) calibration laboratory accredited by A2LA™; indicates traceability to NIST™.
Specifications
Specifications
ISO 17025, A2LA, NIST
3.56 mm
Traceable certificate, Stainless steel probe, Protective sleeve
280 mm
Plastic
11 in.
150°C
-50°C
-50°C to +150°C
Digital LCD
0.14 in.
Measuring temperature in soil, cuvettes, test tubes, food products, incubators and petroleum hydrocarbons
8 in.
20 cm
0.1°C
302°F
-58°F
-58°F to +302°F
±0.2°C
Videos
SDS
Documents
Documents
Accessories