missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Isotemp™ General Purpose Heating and Drying Ovens

Consistent, reliable results

Supplier:  Fisherbrand™ 51030509

 View more versions of this product

Catalog No. 15-103-0509


Explore available promotions

Description

Description

Consistent, reliable results

  • Outstanding temperature uniformity and stability to keep samples at desired condition
  • Wide temperature range: 50° to 250°C
  • Intuitive and easy–to–use microprocessor controller
  • Large, easy–to–read fluorescent display
  • Automatic over temperature alarm
  • On/Off timer for automated operation
  • Easy calibration routine ensures temperature compliance over time
  • Corrosion resistant stainless steel interior (1.4016 / AISI 430) with smooth corners for easy cleaning
  • Manual controlled fresh air damper to adjust air exchange
  • Doors can open to over 180° degrees for optimal access
  • Flexible shelving system with different positions to optimize chamber space
  • Outstanding footprint/volume ratio reduces required bench space
  • Stackable same–size–units, with optional heat shield, without the need for tools
  • Stainless steel exterior provides a clean, professional look for your lab and the surface is structured to avoid showing fingerprints

Applications:

Sample drying, baking, aging tests, glassware drying, dry sterilization, processing electronics

Certifications

cCSAus certified

Warranty

Two year warranty on parts and labor

Specifications

Specifications

6.2 cu.ft.
176 L
Gravity Convection
2 supplied/19 max.
±0.4°C
23.2 x 17.2 x 26.8 in. (589 x 438 x 680 mm)
120 V 60 Hz 1680 w 14 A Nema 5-15 US
1680 w
60 Hz
cCSAus
50°C to 250°C
Gravity convection
Nema 5-15 US
±5°C
29.1 x 25.2 x 36.2 in. (738 x 640 x 920 mm)
120 V
14 A
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions