missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ Model 120 Sonic Dismembrator

Programable system ideal for small volume liquid processing. There are 3 probe size options available which allow for processing 0.2-50 mL volumes.

Supplier:  Fisherbrand™ FB120110

 View more versions of this product

Catalog No. FB120110


Explore available promotions

Includes: Generator, converter, 1/8 in. probe for processing 0.5-15ml volumes, cables, wrench set.

Description

Description

Use this sonic dismembrator for effective analysis and sample preparation of small sample sizes. Fisher Scientific™ Model 120 Sonic Dismembrator offers both manual and programmable operation for consistent and reproducible results. Processes 0.2 to 50mL.

  • Processes 0.2 to 50mL
  • 120 watts
  • Programmable from 1 second to 10 hours
  • Overload protection
  • Pulse mode (1 second to 59 seconds)
  • Energy Display(Watts and Joules)
  • Available accessories include: 3 Probe sizes for individual samples, Eight tip horn for increased sample throughput, Cup Horn for indirect sonication
  • Lightweight converter for handheld operation

Order Info

Stand not included.

Specifications

Specifications

110/117 V
20 kHz (Ultrasonic Frequency)
0.2 to 50mL
6 in.
13.5 in.
8 in.
Cell disruption, protein extraction, DNA shearing/ChIP assay
FB120
110/117 V, 50/60 Hz
120 W
13.5 x 8 x 6 in. (34.29 x 20.32 x 15.24 cm)
15.24 cm
34.29 cm
20.32 cm
Generator, converter, 1/8 in. probe, power cable, wrench set. Note: Stand and Clamp shown in picture not included
1 sec. to 10 hrs.
Videos
SDS
Documents

Documents

Accessories
Promotions

Promotions