missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fisherbrand™ MultiTube Vortexers: Test Tube Racks
Test tube racks for Fisherbrand™ MultiTube Vortexers
Supplier: Fisherbrand™ 80862905
Description
Test tube racks for Fisherbrand™ MultiTube Vortexers
For use with Fisher Scientific MultiTube Vortexers
Color-coded foam racks for test tubes
Specifications
Tubes | |
Test Tube Rack | |
Fisher Scientific MultiTube Vortexer |
25 mm | |
White | |
28 |