missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fisherbrand™ MultiTube Vortexers: Test Tube Racks

Test tube racks for Fisherbrand™ MultiTube Vortexers

$81.42 - $126.42

Specifications

Product Type Test Tube Rack
For Use With (Equipment) Fisher Scientific MultiTube Vortexer
View More Specs

Products 4
Catalog Number Mfr. No. Tube Size Color Price Quantity  
Catalog Number Mfr. No. Tube Size Color Price Quantity  
02-215-454
View Documents
Fisherbrand™
80862901
100 mL Gray
Each for $81.42
 
02-215-455
View Documents
Fisherbrand™
80862902
12 mm Blue
Each for $86.24
 
02-215-456
View Documents
Fisherbrand™
80862903
13 mm Yellow
Each for $84.18
 
02-215-458
View Documents
Fisherbrand™
80862905
25 mm White
Each for $126.42
 
Description

Description

Test tube racks for Fisherbrand™ MultiTube Vortexers

For use with Fisher Scientific MultiTube Vortexers

Color-coded foam racks for test tubes

Specifications

Specifications

Test Tube Rack
Fisher Scientific MultiTube Vortexer
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'