Fisherbrand™ Stainless-Steel Lab Lifts
Over-sized, sure-grip adjustment knobs provide extra leverage for easy height adjustments.
 View more versions of this product
Catalog No. 02-216-477
Explore available promotions
Description
Description
Over-sized, sure-grip adjustment knobs provide extra leverage for easy height adjustments.
  • Ultra-stable lifting platforms
  • Chemical resistant
  • Autoclavable
  • Hold variety of items such as glassware, hotplates, baths and magnetic stirrers
  • Suited for use in fume hoods or bench tops
  • Can withstand extreme environments and high load applications
  • Available in seven convenient sizes
Specifications
Specifications
Stainless Steel
3 to 9.755 in.
133 lb.
Autoclavable
7.6 to 24.8 cm
6 x 6 in.
Lab lift
Product Suggestions
Product Suggestions