Fisherbrand™ Stainless-Steel Lab Lifts
Over-sized, sure-grip adjustment knobs provide extra leverage for easy height adjustments.
Description
Over-sized, sure-grip adjustment knobs provide extra leverage for easy height adjustments.
- Ultra-stable lifting platforms
- Chemical resistant
- Autoclavable
- Hold variety of items such as glassware, hotplates, baths and magnetic stirrers
- Suited for use in fume hoods or bench tops
- Can withstand extreme environments and high load applications
- Available in seven convenient sizes
Specifications
Stainless Steel | |
3.5 to 13 in. | |
227 lb. | |
Autoclavable |
8.9 to 33 cm | |
8 x 8 in. | |
Lab lift | |
1 Ea. |