Fisherbrand™ Stainless-Steel Lab Lifts
Over-sized, sure-grip adjustment knobs provide extra leverage for easy height adjustments.
 View more versions of this product
Catalog No. 02-216-480
Explore available promotions
Description
Description
Over-sized, sure-grip adjustment knobs provide extra leverage for easy height adjustments.
  • Ultra-stable lifting platforms
  • Chemical resistant
  • Autoclavable
  • Hold variety of items such as glassware, hotplates, baths and magnetic stirrers
  • Suited for use in fume hoods or bench tops
  • Can withstand extreme environments and high load applications
  • Available in seven convenient sizes
Specifications
Specifications
Stainless Steel
4 to 19.5 in.
100 lb.
Autoclavable
10.2 to 49.5 cm
12 x 12 in.
Lab lift
Product Suggestions
Product Suggestions