Fisherbrand™ Test Tube Holders for Heavy-duty Vortex Mixer
For use with heavy-duty vortex mixer.
Supplier:  Fisherbrand™ 80863051
 View more versions of this product
Catalog No. 02-216-125
Explore available promotions
Description
Description
  • Holds a wide range of test tube numbers and diameters
  • Requires Universal Holder
Specifications
Specifications
Test Tube Holder
Heavy-duty vortex mixer
34
12 to 13 mm
Foam insert
Universal holder
Videos
SDS
Documents
Documents
Accessories