Fisherbrand™ Disposable Base Molds Edge
Edge
For use in the preparation of pathology specimens
 View more versions of this product
Catalog No. 22-363-553
Edge
Explore available promotions
Description
Description
For use in the preparation of pathology specimens
  • Available in common base mold sizes; can be used with the same styles and types of cassettes and embedding rings
  • Molds are made from specially formulated plastic, offering excellent thermal exchange
  • A smooth interior finish and round corners facilitate specimen removal
Specifications
Specifications
Base Mold
15 mm
0.6 in.
0.5 cm
0.6 x 0.6 x 0.2 in. (15 x 15 x 5 mm)
Can be use with same styles and types of cassettes and embedding rings and provides offering excellent thermal exchange
Plastic
15 mm
0.6 in.
0.19 in.
Single-use
500/Pk.