Fisherbrand™ Tissue Path™ Disposable Embedding Rings
Tissue Path™ Disposable Embedding Rings fit both shallow and deep base molds.
Description
- High-impact polystyrene ensures accurate positioning of tissue samples
- Etched marking surface can be easily written on for positive sample identification
- Rings fit both shallow and deep base molds
Order Info
Packaged in four dispenser boxes of 250.
Specifications
Embedding Ring | |
Green | |
Designed for use with both shallow and deep base molds, for holding and identifying tissue sample blocks and fit well in microtome chuck adapters |
Polystyrene | |
Single-use | |
250 Pk. |