Fisherbrand™ Traceable™ Handheld Digital Barometer
Keep track of your environment and your time with this multi-functional Traceable™ Handheld Digital Barometer.
Description
Keep track of your environment and your time with this multi-functional Traceable™ Handheld Digital Barometer.
- Stopwatch/time of day clock
- Barometric pressure trend displays current reading and previous 2, 4, 6, 8, 10 and 12 hours in a bar chart format
- Minimum/maximum memory for altitudes reached during any time period
- Barometer Accuracy: ±8 mbar (0.2363 in. Hg)
Certifications
Traceable to standards provided by NIST™. Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA.
Specifications
-10°C to +60°C | |
0.1°C | |
Barometer, Lanyard, Two Button-cell Batteries, Shock-resistant Case, No Battery |
±8 mbar | |
14°F to 140°F | |
Handheld Digital Barometer |